SY3H3D67C6D9 Synthonix Corporation

4-(3,5-Difluorophenyl)tetrahydro-2H-pyran-4-ol

  • Cas Number : 139503-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54567.html

Safety Information

MSDS https://synthonix.com/54567.html
Availability details for
Quantity
  • Availability
  • Add to Cart