SY3H3D67C6E0 Synthonix Corporation

N-(4-(2-(3,5-Diamino-4H-pyrazol-4-ylidene)hydrazinyl)phenyl)acetamide

  • MDL number MFCD25972930
  • Cas Number : 333326-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54628.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54628.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart