SY3H3D67C6ED Synthonix Corporation

2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD09037475
  • Cas Number : 1072945-00-6
  • Molecular Weight : 241.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54738.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54738.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart