SY3H3D67C6FB Synthonix Corporation

2,6-Dimethyl-4-(5-nitropyridin-2-yl)morpholine

  • MDL number MFCD00118337
  • Cas Number : 260447-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54843.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54843.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart