SY3H3D67CBD0 Synthonix Corporation

(2,6-Dimethylphenyl)(1H-imidazol-4-yl)methanol

  • MDL number MFCD01721417
  • Cas Number : 78892-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55863.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55863.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart