SY3H3D67CBD1 Synthonix Corporation

3-(1-(Dimethylamino)ethyl)phenyl dimethylcarbamate hydrochloride

  • Cas Number : 105601-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57448.html

Safety Information

MSDS https://synthonix.com/57448.html
Availability details for
Quantity
  • Availability
  • Add to Cart