SY3H3D67CBD3 Synthonix Corporation

5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride

  • Cas Number : 120013-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59105.html

Safety Information

MSDS https://synthonix.com/59105.html
Availability details for
Quantity
  • Availability
  • Add to Cart