SY3H3D67CBD4 Synthonix Corporation

2,2-Dibutoxyethyl ether

  • MDL number MFCD00009459
  • Cas Number : 112-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Almost colorless
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59608.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59608.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart