SY3H3D67CBD8 Synthonix Corporation

Ethyl 3-oxopiperidine-4-carboxylate hydrochloride

  • MDL number MFCD12828687
  • Cas Number : 72738-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49554.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49554.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart