SY3H3D67CBE1 Synthonix Corporation

Diethyl 2-isopropyl-3-oxosuccinate

  • MDL number MFCD00511085
  • Cas Number : 59916-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55031.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55031.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart