SY3H3D67CC05 Synthonix Corporation

(3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methanamine hydrochloride

  • MDL number MFCD28038688
  • Cas Number : 35202-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55513.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55513.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart