SY3H3D67CC0B Synthonix Corporation

3,5-Di(pyridin-4-yl)-4H-1,2,4-triazol-4-amine

  • Cas Number : 38634-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55572.html

Safety Information

MSDS https://synthonix.com/55572.html
Availability details for
Quantity
  • Availability
  • Add to Cart