SY3H3D67CC0C Synthonix Corporation

6,7-Dimethoxy-4-(trifluoromethyl)quinolin-2(1H)-one

  • MDL number MFCD06654703
  • Cas Number : 249736-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55591.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55591.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart