SY3H3D67CC0D Synthonix Corporation

3,3-Dimethyl-3,4-dihydro-1H-furo[3,4-b]indol-1-one

  • MDL number MFCD25460335
  • Cas Number : 139927-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55670.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55670.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart