SY3H3D67CC0F Synthonix Corporation

3-(1,3-Dioxo-3,4-dihydroisoquinolin-2(1H)-yl)propanoic acid

  • MDL number MFCD07310140
  • Cas Number : 75513-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55688.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55688.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart