SY3H3D67CC15 Synthonix Corporation

1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00020785
  • Cas Number : 83-10-3
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white to pale-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55723.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55723.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart