SY3H3D67CDA1 Synthonix Corporation

(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol

  • MDL number MFCD28902278
  • Cas Number : 133775-25-4
  • Molecular Weight : 269.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55803.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55803.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart