SY3H3D67CDA5 Synthonix Corporation

3-((Dimethylamino)methyl)-4-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD03011572
  • Cas Number : 87743-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54894.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54894.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart