SY3H3D67CDAC Synthonix Corporation

2-(3,5-Dimethoxyphenyl)-2-methylpropanenitrile

  • MDL number MFCD09749864
  • Cas Number : 22972-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55981.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55981.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart