SY3H3D67CDCD Synthonix Corporation

2-(3,3-Dimethylbut-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD08276881
  • Cas Number : 154820-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56326.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56326.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart