SY3H3D67CDD1 Synthonix Corporation

Dicyclopentyldimethoxysilane

  • MDL number MFCD01074471
  • Cas Number : 126990-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56345.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/56345.html
pictograms GHS07
hstatements H227, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart