SY3H3D67CDD6 Synthonix Corporation

4,6-Dibromodibenzo[b,d]thiophene

  • MDL number MFCD18451473
  • Cas Number : 669773-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56364.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56364.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart