SY3H3D67CDD7 Synthonix Corporation

2,9-Dibromo-1,10-phenanthroline

  • Cas Number : 39069-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/56365.html

Safety Information

MSDS https://synthonix.com/56365.html
Availability details for
Quantity
  • Availability
  • Add to Cart