SY3H3D67CDDE Synthonix Corporation

2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine

  • Cas Number : 387827-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56377.html

Safety Information

MSDS https://synthonix.com/56377.html
Availability details for
Quantity
  • Availability
  • Add to Cart