SY3H3D67CDDF Synthonix Corporation

4,6-Dibromodibenzo[b,d]furan

  • Cas Number : 201138-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56380.html

Safety Information

MSDS https://synthonix.com/56380.html
Availability details for
Quantity
  • Availability
  • Add to Cart