SY3H3D67CDE3 Synthonix Corporation

1,2-Di(pyridin-4-yl)ethyne

  • Cas Number : 73564-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56413.html

Safety Information

MSDS https://synthonix.com/56413.html
Availability details for
Quantity
  • Availability
  • Add to Cart