SY3H3D67CDE5 Synthonix Corporation

(4,6-Dichloropyrimidin-5-yl)(4-methoxyphenyl)methanone

  • MDL number MFCD11109887
  • Cas Number : 1245646-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56419.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56419.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart