SY3H3D67CDEB Synthonix Corporation

(3',4'-Dichloro-[1,1'-biphenyl]-4-yl)boronic acid

  • Cas Number : 1025736-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56530.html

Safety Information

MSDS https://synthonix.com/56530.html
Availability details for
Quantity
  • Availability
  • Add to Cart