SY3H3D67CE45 Synthonix Corporation

5-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-1H-pyrrole-2-carboxylic acid

  • Cas Number : 1225733-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56724.html

Safety Information

MSDS https://synthonix.com/56724.html
Availability details for
Quantity
  • Availability
  • Add to Cart