SY3H3D67CE4A Synthonix Corporation

(E)-3,5-Dichloro-4-(3-ethoxy-2-methyl-3-oxoprop-1-en-1-yl)benzoic acid

  • Cas Number : 1110767-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56794.html

Safety Information

MSDS https://synthonix.com/56794.html
Availability details for
Quantity
  • Availability
  • Add to Cart