SY3H3D67CE5E Synthonix Corporation

Diethyl 2-(phenylamino)malonate

  • MDL number MFCD00122281
  • Cas Number : 6414-58-0
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2-anilinomalonate
Inchi Code 1S/C13H17NO4/c1-3-17-12(15)11(13(16)18-4-2)14-10-8-6-5-7-9-10/h5-9,11,14H,3-4H2,1-2H3
Inchi Key RUEYBBPBTGHLNO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/57039.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57039.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart