SY3H3D67CE64 Synthonix Corporation

2,6-Diisopropylbenzoic Acid

  • MDL number No MDL found
  • Cas Number : 92035-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57113.html

Safety Information

MSDS https://synthonix.com/57113.html
Availability details for
Quantity
  • Availability
  • Add to Cart