SY3H3D67CE78 Synthonix Corporation

2,7-Dichloro-9H-fluorene

  • MDL number MFCD00032840
  • Cas Number : 7012-16-0
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57308.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57308.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart