SY3H3D67CE81 Synthonix Corporation

2-(2,6-Dioxopiperidin-3-yl)-4-nitroisoindoline-1,3-dione

  • MDL number MFCD01748356
  • Cas Number : 19171-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57361.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57361.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart