SY3H3D67CE82 Synthonix Corporation

1-(3,4-Difluorobenzyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD11111121
  • Cas Number : 1001413-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57369.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57369.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart