SY3H3D67CE86 Synthonix Corporation

10,11-Dihydro-11-oxodibenzo[b,f][1,4]thiazepine

  • MDL number MFCD00901197
  • Molecular Weight : 227.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57377.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57377.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart