SY3H3D67CEE0 Synthonix Corporation

Dimethyl [2,2'-bipyridine]-3,3'-dicarboxylate

  • Cas Number : 39775-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57558.html

Safety Information

MSDS https://synthonix.com/57558.html
Availability details for
Quantity
  • Availability
  • Add to Cart