SY3H3D67CEE4 Synthonix Corporation

Diethyl dithieno[3,2-b:2',3'-d]thiophene-2,6-dicarboxylate

  • Cas Number : 502764-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57585.html

Safety Information

MSDS https://synthonix.com/57585.html
Availability details for
Quantity
  • Availability
  • Add to Cart