SY3H3D67CEE8 Synthonix Corporation

(2,4-Dimethylphenyl)(2-nitrophenyl)sulfane

  • MDL number MFCD27918698
  • Cas Number : 1610527-49-5
  • Molecular Weight : 259.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dimethylphenyl)(2-nitrophenyl)sulfane
Inchi Code 1S/C14H13NO2S/c1-10-7-8-13(11(2)9-10)18-14-6-4-3-5-12(14)15(16)17/h3-9H,1-2H3
Inchi Key SHBWPKLGXVSPIP-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/57620.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/57620.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart