SY3H3D67CEF5 Synthonix Corporation

Diethyl 2-(2-bromo-4-nitrophenyl)-2-methylmalonate

  • MDL number MFCD28902467
  • Cas Number : 945244-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57720.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57720.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart