SY3H3D67CEF7 Synthonix Corporation

1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione

  • MDL number MFCD23703120
  • Cas Number : 1231160-83-0
  • Molecular Weight : 423.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57737.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57737.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart