SY3H3D67CF04 Synthonix Corporation

2-(2,3-Dihydrobenzofuran-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD10699453
  • Cas Number : 445303-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57870.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57870.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart