SY3H3D67CF0A Synthonix Corporation

2-(2,5-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD12405521
  • Cas Number : 1073339-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57939.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57939.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart