SY3H3D67CF15 Synthonix Corporation

N-(2-(Diethylamino)ethyl)-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxamide

  • Cas Number : 356068-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/57966.html

Safety Information

MSDS https://synthonix.com/57966.html
Availability details for
Quantity
  • Availability
  • Add to Cart