SY3H3D67CF16 Synthonix Corporation

N-(3,5-Dimethyladamantan-1-yl)acetamide

  • Cas Number : 19982-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57969.html

Safety Information

MSDS https://synthonix.com/57969.html
Availability details for
Quantity
  • Availability
  • Add to Cart