SY3H3D67CF71 Synthonix Corporation

N,N-Dihexylacetamide

  • MDL number MFCD00082658
  • Cas Number : 16423-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to pale-yellow Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58010.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58010.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart