SY3H3D67CF7D Synthonix Corporation

N,N-Dimethylpyridin-4-amine 4-methylbenzenesulfonate

  • MDL number MFCD25372259
  • Cas Number : 91944-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58072.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58072.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart