SY3H3D67CF84 Synthonix Corporation

2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one hydrate

  • MDL number MFCD04220857
  • Cas Number : 1174865-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58126.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58126.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart