SY3H3D67CF88 Synthonix Corporation

2-((Diphenylmethylene)amino)acetonitrile

  • MDL number MFCD00009970
  • Cas Number : 70591-20-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58149.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58149.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart