SY3H3D67CF8A Synthonix Corporation

2,7-Dibromo-9,9-dimethyl-9,10-dihydroacridine

  • MDL number MFCD26402967
  • Cas Number : 1333316-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58184.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58184.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart